Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhodamine 6G

Supplier: TCI America
Synonyms: 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

R0039-25G
TCR0039-025GEA 57.21 USD InStock
TCR0039-025G
Rhodamine 6G
Rhodamine 6G
CAS Number: 989-38-8
MDL Number: MFCD00012665
Molecular Formula: C28H31ClN2O3
Molecular Weight: 479.02
Form: Crystal
Color: Deep Yellow Red

Other Product Names / Synonyms: Basic Red 1


Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/56/28993756.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty