Would you like to log in again?
NeuLog Salinity Logger Sensor
NUL-228
470137-422EA 130 USD InStock
470137-422
NeuLog Salinity Logger Sensor
Electrochemical Meters Salinity Meters
This logger sensor quickly measures salt content.
  • Three Measurable Ranges: %, mg/L, and ppm
  • Sensor Supports Experiments Lasting from One Second to 31 Days
  • Sensor Measures Quickly
Measure salt content of solutions in three ranges: %, mg/L, and ppm.
This sensor supports experiments that last from 1 second to 31 days.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty