Would you like to log in again?
NeuLog Colorimeter Logger Sensor
Supplier: EISCO SCIENTIFIC LLC
NUL-219
470137-406EA 175.05 USD InStock
470137-406
NeuLog Colorimeter Logger Sensor
Data Loggers
Use for Beer's Law experiments.
  • Max. sample rate 100 S/sec.
The colorimeter measures the levels of color components (red, green and blue) of a solution to determine concentration. It can also be used to determine the solution absorbance of each color component. This sensor has two modes of operation: Absorption and Transparency.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module(Not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty