Synonyms:
Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate
, Food Blue No.1
, Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonate
, Erioglaucine sodium salt
, Alphazurine FG
, CI 42090
, Erioglaucin A
, Acid blue 9
, FD and C Blue No. 1
, Brilliant Blue FCF
B0790-25G
TCB0790-025GEA
64.35
USD
InStock
TCB0790-025G
Erioglaucine disodium salt
Erioglaucine disodium salt
CAS Number: 3844-45-9
MDL Number: MFCD00012141
Molecular Formula: C37H36N2O9S3
Molecular Weight: 792.84
Form: Crystal
Color: Red
Other Product Names / Synonyms: Brilliant Blue FCF, Erioglaucin A