Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bisabolene (so called)

Supplier: TCI America
Synonyms: 4-(1,5-Dimethyl-4-hexenylidene)-1-methylcyclohexene

B1413-25ML
TCB1413-025MLEA 93.33 USD InStock
TCB1413-025ML
Bisabolene (so called)
Bisabolene
[for Fragrance]
MDL Number: MFCD00129080
Molecular Formula: C15H24
Molecular Weight: 204.36
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 200


Formula: C₁₅H₂₄
MW: 204.36 g/mol
Flash Pt: 200 °C
MDL Number: MFCD00129080
CAS Number: 495-62-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty