Synonyms:
3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid
, 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid
, 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulfonic acid
, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
B1494-25G
TCB1494-025GEA
80.7
USD
InStock
TCB1494-025G
DIPSO ≥98.0% (by HPLC, titration analysis)
DIPSO
CAS Number: 68399-80-4
MDL Number: MFCD00038353
Molecular Formula: C7H17NO6S
Molecular Weight: 243.27
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: DIPSO