Synonyms:
p,p'-Dicarboxy-1,10-diphenoxydecane
B1499-5G
TCB1499-005GEA
1564
USD
InStock
TCB1499-005G
1,10-Bis(4-carboxyphenoxy)decane ≥97.0% (by titrimetric analysis)
1,10-Bis(4-carboxyphenoxy)decane
CAS Number: 74774-61-1
MDL Number: MFCD00142356
Molecular Formula: C24H30O6
Molecular Weight: 414.50
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 274
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: p,p'-Dicarboxy-1,10-diphenoxydecane