Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol ≥96.0%

Supplier: TCI America
Synonyms: 1,2-Diphenyl-1,2-di(p-tolyl)-1,2-ethanediol

B1849-1G
TCB1849-001GEA 164.35 USD InStock
TCB1849-001G
1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol ≥96.0%
1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol
CAS Number: 808-12-8
MDL Number: MFCD00026012
Molecular Formula: C28H26O2
Molecular Weight: 394.51
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 156

Other Product Names / Synonyms: 1,2-Diphenyl-1,2-di(p-tolyl)-1,2-ethanediol


Formula: C₂₈H₂₆O₂
MW: 394.51 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00026012
CAS Number: 808-12-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/36/15/28933615.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty