Synonyms:
Arecoline hydrobromide
A0523-5G
A0523-25G
TCA0523-025GEA
255.78
USD
InStock
TCA0523-025G
TCA0523-005G
Arecolinium bromide ≥98.0% (by HPLC, titration analysis)
Arecolinium bromide
CAS Number: 300-08-3
MDL Number: MFCD00039041
Molecular Formula: C8H13NO2
Molecular Weight: 236.11
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 172