Synonyms:
4,4'-Cyclohexylidenebis(o-cresol)
, 4,4'-Cyclohexylidenebis(2-methylphenol)
B2753-25G
TCB2753-25GEA
136
USD
InStock
TCB2753-25G
1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane ≥98.0%
1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane
CAS Number: 2362-14-3
Molecular Formula: C20H24O2
Molecular Weight: 296.41
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 191
Product Categories: Materials Chemistry, Monomers, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4,4'-Cyclohexylidenebis(o-cresol), 4,4'-Cyclohexylidenebis(2-methylphenol)