B3367-5G
B3367-25G
TCB3367-25GEA
167.25
USD
InStock
TCB3367-25G
TCB3367-5G
α-Bromo-4-chlorophenylacetic acid ≥98.0% (by titrimetric analysis)
α-Bromo-4-chlorophenylacetic acid
CAS Number: 3381-73-5
MDL Number: MFCD08276760
Molecular Formula: C8H6BrClO2
Molecular Weight: 249.49
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 98