Synonyms:
(S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol
C0005-1G
TCC0005-001GEA
75.37
USD
InStock
TCC0005-001G
Ergocalciferol (vitamin D2, calciferol) ≥98.0% (by HPLC)
Ergocalciferol (Vitamin D2, Calciferol)
CAS Number: 50-14-6
MDL Number: MFCD00166988
Molecular Formula: C28H44O
Molecular Weight: 396.66
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 114
Specific rotation [a]20/D: 105 deg (C=1.5, EtOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Anti-osteoporosis Ingredients, Vitamins
Other Product Names / Synonyms: Ergocalciferol, Vitamin D2