Synonyms:
(2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride
, (2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid) dihydrochloride
C0520-25G
TCC0520-025GEA
49.72
USD
InStock
TCC0520-025G
L-Cystine dihydrochloride ≥98.0% (by HPLC)
L-Cystine dihydrochloride
CAS Number: 30925-07-6
MDL Number: MFCD00070399
Molecular Formula: C6H12N2O4S2
Molecular Weight: 313.21
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -168 deg (C=1, 1mol/L HCl)
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: (H-Cys-OH)2.2HCl