Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chrysophenine

Supplier: TCI America
Synonyms: Chrysophenine sodium salt

C0530-500G C0530-25G
TCC0530-500GEA 194.67 USD InStock
TCC0530-500G TCC0530-025G
Chrysophenine
Chrysophenine
CAS Number: 2870-32-8
MDL Number: MFCD00007488
Molecular Formula: C30H28N4O8S2
Molecular Weight: 680.66
Form: Crystal
Color: Yellow


Formula: C₃₀H₂₈N₄O₈S₂
MDL Number: MFCD00007488
CAS Number: 2870-32-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty