Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nε-carbobenzoxy-Nɑ-tosyl-L-lysine ≥98.0% (by HPLC)

Supplier: TCI America
Synonyms: Nε-Cbz-Nɑ-tosyl-L-lysineTos-Lys(Z)-OH

C0735-5G C0735-1G
TCC0735-001GEA 96 USD InStock
TCC0735-001G TCC0735-5G
Nε-carbobenzoxy-Nɑ-tosyl-L-lysine ≥98.0% (by HPLC)
Nepsilon-carbobenzoxy-Nalpha-tosyl-L-lysine
CAS Number: 34235-82-0
MDL Number: MFCD00059201
Molecular Formula: C21H26N2O6S
Molecular Weight: 434.51
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 123
Specific rotation [a]20/D: 14.5 deg (C=1, EtOH)

Other Product Names / Synonyms: Nepsilon-Cbz-Nalpha-tosyl-L-lysine, Tos-Lys(Z)-OH


Formula: C₂₁H₂₆N₂O₆S
MW: 434.51 g/mol
Melting Pt: 123 °C
MDL Number: MFCD00059201
CAS Number: 34235-82-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/80/28937780.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty