Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropyl phenyl ketone ≥98.0%

Supplier: TCI America
Synonyms: BenzoylcyclopropaneCyclopropyl(phenyl)methanone

C0987-25ML
TCC0987-025MLEA 132.05 USD InStock
TCC0987-025ML
Cyclopropyl phenyl ketone ≥98.0%
Cyclopropyl phenyl ketone
CAS Number: 3481-02-5
MDL Number: MFCD00001294
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Flash Point (°C): 90
Specific Gravity (20/20): 1.06

Other Product Names / Synonyms: Benzoylcyclopropane


Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 248 °C
Density: 1.06 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00001294
CAS Number: 3481-02-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/82/41/28938241.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty