C1666-5G
C1666-25G
TCC1666-25GEA
264
USD
InStock
TCC1666-25G
TCC1666-5G
5-Chloro-2,4,6-trifluoropyrimidine ≥98.0%
5-Chloro-2,4,6-trifluoropyrimidine
CAS Number: 697-83-6
MDL Number: MFCD00191933
Molecular Formula: C4ClF3N2
Molecular Weight: 168.50
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.63