Synonyms:
α-CHCA
, 2-Cyano-3-(4-hydroxyphenyl)acrylic Acid
, α-Cyano-4-hydroxycinnamic Acid
, CHCA
C1768-1G
TCC1768-1GEA
136.2
USD
InStock
TCC1768-1G
ɑ-Cyano-4-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)
alpha-Cyano-4-hydroxycinnamic acid
CAS Number: 28166-41-8
MDL Number: MFCD00004204
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 250
Product Categories: Bioscience, Phenylpropanoids, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2-Cyano-3-(4-hydroxyphenyl)acrylic Acid, alpha-CHCA