Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iota carrageenan

Supplier: TCI America

C1805-500G C1805-25G
TCC1805-500GEA 164.13 USD InStock
TCC1805-500G TCC1805-25G
Iota carrageenan
Iota carrageenan
CAS Number: 9062-07-1
MDL Number: MFCD00151512
Form: Crystal
Color: White

Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)


MDL Number: MFCD00151512
CAS Number: 9062-07-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/90/28938990.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty