Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nɑ-Carbobenzoxy-D-arginine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: Nɑ-Cbz-D-arginineZ-D-Arg-OH

C2131-5G
TCC2131-5GEA 415 USD InStock
TCC2131-5G
Nɑ-Carbobenzoxy-D-arginine ≥98.0% (by HPLC, titration analysis)
Nalpha-Carbobenzoxy-D-arginine
CAS Number: 6382-93-0
MDL Number: MFCD00063009
Molecular Formula: C14H20N4O4
Molecular Weight: 308.34
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175
Specific rotation [a]20/D: 8 deg (C=5.5, 0.2mol/L HCl)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-D-arginine, Z-D-Arg-OH


Formula: C₁₄H₂₀N₄O₄
MW: 308.34 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00063009
CAS Number: 6382-93-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/79/28939179.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty