Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Cysteine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: H-D-Cys-OMe.HCl

C2174-25G C2174-1G C2174-5G
TCC2174-25GEA 420.09 USD InStock
TCC2174-25G TCC2174-1G TCC2174-5G
D-Cysteine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
D-Cysteine methyl ester hydrochloride
CAS Number: 70361-61-4
MDL Number: MFCD00173915
Molecular Formula: C4H9NO2S
Molecular Weight: 171.64
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 135
Specific rotation [a]20/D: 3 deg (C=20, MeOH)

Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Cys-OMe.HCl


Formula: C₄H₉NO₂S
MW: 171.64 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00173915
CAS Number: 70361-61-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/95/28939195.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty