Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: meso-1,2-Bis(4-pyridyl) Glycolmeso-1,2-Di(4-pyridyl)-1,2-ethanediolmeso-1,2-Di(4-pyridyl) Glycol

D0936-5G
TCD0936-005GEA 194.82 USD InStock
TCD0936-005G
meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)
meso-alpha,beta-Di(4-pyridyl) glycol
CAS Number: 4972-49-0
MDL Number: MFCD00023627
Molecular Formula: C12H12N2O2
Molecular Weight: 216.24
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Other Product Names / Synonyms: meso-1,2-Bis(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl)-1,2-ethanediol


Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
MDL Number: MFCD00023627
CAS Number: 4972-49-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/83/28976083.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty