Synonyms:
3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid
, 3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid
, 3,5-Diacetamido-2,4,6-triiodobenzoic acid
D1462-25G
TCD1462-025GEA
73.88
USD
InStock
TCD1462-025G
Diatrizoic acid ≥98.0% (by HPLC, titration analysis)
Diatrizoic acid
CAS Number: 117-96-4
MDL Number: MFCD00069960
Molecular Formula: C11H9I3N2O4
Molecular Weight: 613.92
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid