D2300-5G
TCD2300-005GEA
262.76
USD
InStock
TCD2300-005G
1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine ≥92.0% (by titrimetric analysis)
1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine
CAS Number: 22429-12-5
MDL Number: MFCD00043134
Molecular Formula: C10H15N2P
Molecular Weight: 194.22
Purity/Analysis Method: 92.0% (T)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 108
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C