Synonyms:
(+)-2,3:5,6-Di-O-isopropylidene-α-D-mannofuranoseD-Mannose diacetonide0
D2447-5G
TCD2447-005GEA
56
USD
InStock
TCD2447-005G
2,3:5,6-Di-O-isopropylidene-D-mannofuranose ≥98.0%
2,3:5,6-Di-O-isopropylidene-D-mannofuranose
CAS Number: 14131-84-1
MDL Number: MFCD00134206
Molecular Formula: C12H20O6
Molecular Weight: 260.29
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 125
Specific rotation [a]20/D: 24 deg (C=1, Acetone)