Synonyms:
Borane - N,N-Diethylaniline Complex
D2581-25G
D2581-100G
TCD2581-100GEA
648.23
USD
InStock
TCD2581-100G
TCD2581-025G
N,N-Diethylaniline borane ≥98.0% (by titrimetric analysis)
N,N-Diethylaniline borane
CAS Number: 13289-97-9
MDL Number: MFCD00013187
Molecular Formula: C10H18BN
Molecular Weight: 163.07
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Melting point (°C): -40
Flash Point (°C): 21
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Borane - N,N-Diethylaniline Complex