Synonyms:
Dyphylline
D3600-25G
D3600-500G
TCD3600-25GEA
26.69
USD
InStock
TCD3600-25G
TCD3600-500G
7-(2,3-Dihydroxypropyl)theophylline ≥98.0% (by HPLC)
7-(2,3-Dihydroxypropyl)theophylline
CAS Number: 479-18-5
MDL Number: MFCD00005759
Molecular Formula: C10H14N4O4
Molecular Weight: 254.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 163
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients, Nucleobases and Their Analogs, Purine Alkaloids
Other Product Names / Synonyms: 7-(2,3-Dihydroxypropyl)-1,3-dimethylxanthine, Diprophylline, Dyphylline