Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl orange sodium salt

Supplier: TCI America
Synonyms: 4-Diethylaminoazobenzene-4'-sulfonic Acid Sodium Salt

E0155-25G
TCE0155-025GEA 68.01 USD InStock
TCE0155-025G
Ethyl orange sodium salt
Ethyl orange sodium salt
Form: Crystal
Color: Yellow

Other Product Names / Synonyms: 4-Diethylaminoazobenzene-4'-sulfonic Acid Sodium Salt


Formula: C₁₆H₁₈N₃NaO₃S
MW: 355.39 g/mol
MDL Number: MFCD00007503
CAS Number: 62758-12-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/33/28967733.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty