Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl violet

Supplier: TCI America
Synonyms: Basic Violet 4

E0200-25G
TCE0200-025GEA 36.54 USD InStock
TCE0200-025G
Ethyl violet
Ethyl violet
CAS Number: 2390-59-2
MDL Number: MFCD00011765
Molecular Formula: C31H42ClN3
Molecular Weight: 560.29
Form: Crystal
Color: Deep Yellow Red

Other Product Names / Synonyms: Basic Violet 4


Formula: C₃₁H₄₂ClN₃
MW: 560.29 g/mol
MDL Number: MFCD00011765
CAS Number: 2390-59-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/71/28967771.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty