Synonyms:
4,6-O-Ethylidene-alpha-D-glucose
E0402-5G
E0402-1G
TCE0402-1GEA
65.28
USD
InStock
TCE0402-1G
TCE0402-005G
4,6-O-Ethylidene-ɑ-D-glucopyranose ≥96.0%
4,6-O-Ethylidene-alpha-D-glucopyranose
CAS Number: 13224-99-2
MDL Number: MFCD00006820
Molecular Formula: C8H14O6
Molecular Weight: 206.19
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 173
Product Categories: Bioscience, Glucose, O-Substituted Sugars
Other Product Names / Synonyms: 4,6-O-Ethylidene-alpha-D-glucose