Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorene-2-azo-2',4'-dihydroxybenzene ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 4-(2-Fluorenylazo)resorcinol

F0019-1G
TCF0019-001GEA 117.69 USD InStock
TCF0019-001G
Fluorene-2-azo-2',4'-dihydroxybenzene ≥98.0% (by titrimetric analysis)
Fluorene-2-azo-2',4'-dihydroxybenzene
CAS Number: 63020-83-7
MDL Number: MFCD00059493
Molecular Formula: C19H14N2O2
Molecular Weight: 302.33
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Other Product Names / Synonyms: 4-(2-Fluorenylazo)resorcinol


Formula: C₁₉H₁₄N₂O₂
MW: 302.33 g/mol
MDL Number: MFCD00059493
CAS Number: 63020-83-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/67/28968667.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty