Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erioglaucine disodium salt

Supplier: TCI America
Synonyms: Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonateFood Blue No.1Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonateErioglaucine sodium saltAlphazurine FGCI 42090Erioglaucin AAcid blue 9FD and C Blue No. 1Brilliant Blue FCF

F0147-25G
TCF0147-25GEA 107.18 USD InStock
TCF0147-25G
Erioglaucine disodium salt
Erioglaucine disodium salt
CAS Number: 3844-45-9
MDL Number: MFCD00012141
Molecular Formula: C37H36N2O9S3
Molecular Weight: 792.84
Form: Powder
Color: Blue


Formula: C₃₇H₃₄Na₂N₂O₉S₃
MW: 792.84 g/mol
MDL Number: MFCD00012141
CAS Number: 3844-45-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/64/28968764.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty