Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium formate monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Formic Acid Lithium SaltLithium formate hydrate

F0171-25G F0171-500G
TCF0171-025GEA 47.71 USD InStock
TCF0171-025G TCF0171-500G
Lithium formate monohydrate ≥98.0% (by titrimetric analysis)
Lithium formate monohydrate
CAS Number: 6108-23-2
MDL Number: MFCD00050823
Molecular Formula: CH2O2
Molecular Weight: 51.96
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Other Product Names / Synonyms: Formic Acid Lithium Salt


Formula: HCO₂Li·H₂O
MW: 51.96 g/mol (anhy)
MDL Number: MFCD00050823
CAS Number: 6108-23-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/84/28968784.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty