Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimyristin ≥95.0%

Supplier: TCI America
Synonyms: Glycerol Trimyristate

G0088-25G
TCG0088-025GEA 151.17 USD InStock
TCG0088-025G
Trimyristin ≥95.0%
Trimyristin
CAS Number: 555-45-3
MDL Number: MFCD00036229
Molecular Formula: C45H86O6
Molecular Weight: 723.18
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 57

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Trimyristate


Formula: C₄₅H₈₆O₆
MW: 723.18 g/mol
Melting Pt: 57 °C
MDL Number: MFCD00036229
CAS Number: 555-45-3
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty