Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycyl-DL-Alanine

Supplier: TCI
G0112-1G
TCG0112-001GEA 88.26 USD InStock
TCG0112-001G
Glycyl-DL-Alanine
Proteins and Peptides
  • Melting point: 235 °C
  • EINECS: 213-144-6
  • Purity (HPLC): Min. 98.0 area%
  • Specific rotation [a]20/D: –0.5 to +0.5° (C=8, water)
  • Solubility in water: almost transparency

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-DL-Ala-OH

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty