Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycyl-4-aminobutyric acid

Supplier: TCI America

G0115-1G
TCG0115-001GEA 84.21 USD InStock
TCG0115-001G
Glycyl-4-aminobutyric acid
Glycyl-4-aminobutyric acid
CAS Number: 32595-49-6
MDL Number: MFCD00056123
Molecular Formula: C6H12N2O3
Molecular Weight: 160.17
Form: Crystal
Color: White


Formula: C₆H₁₂N₂O₃
MDL Number: MFCD00056123
CAS Number: 32595-49-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/84/28969784.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty