Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycyl-L-aspartic acid monohydrate

Supplier: TCI
G0121-1G
TCG0121-001GEA 102.52 USD InStock
TCG0121-001G
Glycyl-L-aspartic acid monohydrate
Proteins and Peptides
CAS Number: 4685-12-5
MDL Number: MFCD00025585
Molecular Formula: C6H10N2O5
Molecular Weight: 190.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 13 deg (C=2, H2O)

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Asp-OH

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty