Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexabromostearic acid ≥93.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Hexabromooctadecanoic Acid

H0052-25G
TCH0052-025GEA 200.88 USD InStock
TCH0052-025G
Hexabromostearic acid ≥93.0% (by titrimetric analysis)
Hexabromostearic acid
CAS Number: 4167-08-2
MDL Number: MFCD00021795
Molecular Formula: C18H30Br6O2
Molecular Weight: 757.86
Purity/Analysis Method: 93.0% (T)
Form: Crystal
Melting point (°C): 178

Other Product Names / Synonyms: Hexabromooctadecanoic Acid


Formula: C₁₈H₃₀Br₆O₂
MW: 757.86 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00021795
CAS Number: 4167-08-2
Order Now
Specification Test Results
Purity/Analysis Method 93.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty