Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 2-Hydrazinopyridine2-hydrazinylpyridine2(1H)-Pyridinone, hydrazone

H0888-10G H0888-25G
TCH0888-010GEA 163.85 USD InStock
TCH0888-010G TCH0888-25G
2-Hydrazinylpyridine ≥98.0% (by GC, titration analysis)
2-Hydrazinylpyridine
CAS Number: 4930-98-7
MDL Number: MFCD00006249
Molecular Formula: C5H7N3
Molecular Weight: 109.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 47
Flash Point (°C): 110

Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: 2-Pyridylhydrazine


Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 185 °C
Melting Pt: 47 °C
Flash Pt: 110 °C
MDL Number: MFCD00006249
CAS Number: 4930-98-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/10/28970910.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty