Synonyms:
(S)-1-(Pyridin-2-yl)ethanol
, (S)-1-(2-Pyridyl)ethanol
, (S)-alpha-Methyl-2-pyridinemethanol
H1263-1G
TCH1263-1GEA
120.79
USD
InStock
TCH1263-1G
2-[(S)-1-Hydroxyethyl]pyridine ≥97.0% (by GC, titration analysis)
2-[(S)-1-Hydroxyethyl]pyridine
CAS Number: 59042-90-9
MDL Number: MFCD06795465
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 29
Specific rotation [a]20/D: -29 deg (C=1, Toluene)
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (S)-1-(2-Pyridyl)ethanol, (S)-alpha-Methyl-2-pyridinemethanol