Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium isopropylxanthate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Isopropylxanthic Acid Potassium Salt

I0177-25G
TCI0177-025GEA 165.37 USD InStock
TCI0177-025G
Potassium isopropylxanthate ≥98.0% (by titrimetric analysis)
Potassium isopropylxanthate
CAS Number: 140-92-1
MDL Number: MFCD00004930
Molecular Formula: C4H8OS2
Molecular Weight: 174.32
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Other Product Names / Synonyms: Isopropylxanthic Acid Potassium Salt


Formula: C₄H₈OS₂
MW: 174.32 g/mol
MDL Number: MFCD00004930
CAS Number: 140-92-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/67/28971967.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty