Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2-ethylhexyl)itaconate ≥94.0% stabilized

Supplier: TCI America
Synonyms: Dioctyl ItaconateItaconic Acid Dioctyl EsterItaconic Acid Bis(2-ethylhexyl) Ester

I0205-25ML
TCI0205-025MLEA 209.7 USD InStock
TCI0205-025ML
Bis(2-ethylhexyl)itaconate ≥94.0% stabilized
Bis(2-ethylhexyl)itaconate
(stabilized with HQ)
CAS Number: 2287-83-4
MDL Number: MFCD00059429
Molecular Formula: C21H38O4
Molecular Weight: 354.53
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Flash Point (°C): 187
Specific Gravity (20/20): 0.94

Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Dioctyl Itaconate, Itaconic Acid Dioctyl Ester, Itaconic Acid Bis(2-ethylhexyl) Ester


Formula: C₂₁H₃₈O₄
MW: 354.53 g/mol
Density: 0.94 (20 °C)
Flash Pt: 187 °C
MDL Number: MFCD00059429
CAS Number: 2287-83-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/06/28972006.jpg
Order Now
Specification Test Results
Purity/Analysis Method 94.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty