Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoproterenol hydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Isoprenaline Hydrochloride

I0260-5G I0260-25G
TCI0260-025GEA 296.29 USD InStock
TCI0260-025G TCI0260-5G
Isoproterenol hydrochloride ≥99.0% (by titrimetric analysis)
Isoproterenol hydrochloride
CAS Number: 51-30-9
MDL Number: MFCD00012603
Molecular Formula: C11H17NO3
Molecular Weight: 247.72
Purity/Analysis Method: 99.0% (T)
Form: Crystal

Product Categories: Bioscience, Reagents for Life-style related Disease Models, Regenerative Medicine Research Reagents, Other Pharmaceutical Ingredients, Phenylethylamines
Other Product Names / Synonyms: Isoprenaline Hydrochloride


Formula: C₁₁H₁₇NO₃
MW: 247.72 g/mol
MDL Number: MFCD00012603
CAS Number: 51-30-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/62/28972062.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty