Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isomaltotriose ≥97.0% (by HPLC)

Supplier: TCI America

I0329-1G I0329-100MG
TCI0329-100MGEA 170.82 USD InStock
TCI0329-100MG TCI0329-001G
Isomaltotriose ≥97.0% (by HPLC)
Isomaltotriose
CAS Number: 3371-50-4
MDL Number: MFCD00070623
Molecular Formula: C18H32O16
Molecular Weight: 504.44
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 133 deg (C=1, H2O)


Formula: C₁₈H₃₂O₁₆
MDL Number: MFCD00070623
CAS Number: 3371-50-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/46/28972146.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty