Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-2,4,5-trimethylthiophene ≥96.0% stabilized

Supplier: TCI America

I0445-1G I0445-5G
TCI0445-5GEA 563.08 USD InStock
TCI0445-5G TCI0445-001G
3-Iodo-2,4,5-trimethylthiophene ≥96.0% stabilized
3-Iodo-2,4,5-trimethylthiophene
(stabilized with Copper chip)
CAS Number: 60813-84-5
MDL Number: MFCD00191528
Molecular Formula: C7H9IS
Molecular Weight: 252.11
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Specific Gravity (20/20): 1.68
Storage Temperature: 0-10°C


Formula: C₇H₉IS
MW: 252.11 g/mol
Boiling Pt: 87 °C
Density: 1.68 (20 °C)
MDL Number: MFCD00191528
CAS Number: 60813-84-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/96/28972296.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty