Synonyms:
CPT-11
, (S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate hydrochloride trihydrate
, Irinotecan HCl Trihydrate
I0714-100MG
TCI0714-100MGEA
85.23
USD
InStock
TCI0714-100MG
Irinotecan hydrochloride trihydrate ≥98.0% (by HPLC, titration analysis)
Irinotecan hydrochloride trihydrate
CAS Number: 136572-09-3
MDL Number: MFCD01765731
Molecular Formula: C33H38N4O6
Molecular Weight: 623.15
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 257
Specific rotation [a]20/D: 24 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antitumor Ingredients, Alkaloids (Phytochemicals), Cancer Research Reagents, Quinoline Alkaloids
Other Product Names / Synonyms: CPT-11