Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: Imidazolo[1,5-a]pyridine

I0753-5G
TCI0753-5GEA 172.55 USD InStock
TCI0753-5G
Imidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)
Imidazo[1,5-a]pyridine
CAS Number: 274-47-5
MDL Number: MFCD03407375
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 54
Storage Temperature: 0°C


Formula: C₇H₆N₂
MW: 118.14 g/mol
Melting Pt: 54 °C
MDL Number: MFCD03407375
CAS Number: 274-47-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/26/34/28972634.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty