Synonyms:
4-Iodo-N,N-bis(4-methoxyphenyl)aniline
I0776-5G
I0776-1G
TCI0776-5GEA
318.27
USD
InStock
TCI0776-5G
TCI0776-1G
4-Iodo-4',4''-dimethoxytriphenylamine ≥98.0% (by HPLC, total nitrogen)
4-Iodo-4',4''-dimethoxytriphenylamine
CAS Number: 201802-15-5
Molecular Formula: C20H18INO2
Molecular Weight: 431.27
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 111
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-Iodo-N,N-bis(4-methoxyphenyl)aniline