Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Laminaran (from Eisenia bicyclis)

Supplier: TCI America
Synonyms: β-1,3-Glucan

L0088-1G L0088-25G
TCL0088-025GEA 413.94 USD InStock
TCL0088-025G TCL0088-001G
Laminaran (from Eisenia bicyclis)
Laminaran
CAS Number: 9008-22-4
MDL Number: MFCD00466918
Form: Crystal
Color: White
Specific rotation [a]20/D: -45 deg (C=0.9, H2O)

Product Categories: Bioscience, Natural Polymers (for DDS)


MDL Number: MFCD00466918
CAS Number: 9008-22-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty