Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrichlorosilane ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger
Synonyms: Trichloro(methyl)silane

M0450-100G M0450-500G M0450-25G
TCM0450-500GEA 69.99 USD InStock
TCM0450-500G TCM0450-100G TCM0450-025G
Methyltrichlorosilane ≥98.0% (by GC, titration analysis)
Methyltrichlorosilane
CAS Number: 75-79-6
MDL Number: MFCD00000481
Molecular Formula: CH3Cl3Si
Molecular Weight: 149.47
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 65
Melting point (°C): -90
Flash Point (°C): -8
Specific Gravity (20/20): 1.28

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Methyltrichlorosilane


Formula: CH₃Cl₃Si
MW: 149.48 g/mol
Boiling Pt: 65 °C
Melting Pt: –90 °C
Density: 1.28 (20 °C)
Flash Pt: –8 °C
MDL Number: MFCD00000481
CAS Number: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,I
REACH: 01-2119475226-36
https://us-legacy.vwr.com/stibo/web/std.lang.all/38/90/28973890.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty